Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317338.1 | NP_001304267.1 | 3 | 784 | ||
NM_017898.4 | NP_060368.2 | 3 | 784 | |||
XM_006711407.2 | XP_006711470.1 | 2 | 1340 | |||
XM_011509684.1 | XP_011507986.1 | 3 | 308 | |||
XM_017001581.1 | XP_016857070.1 | 3 | 765 | |||
GenBank mRNA | AL136931.1 | - | 3 | 761 | ||
BC010366.1 | - | 2 | 118 | |||
BC011973.1 | - | 3 | 696 | 3458649 | ||
BC015829.2 | - | 1 | 54 | |||
BC016859.1 | - | 3 | 686 | 3849257 | ||
BP194343.1 | - | 3 | 390 |