Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017905.4 | NP_060375.4 | 7 | 1426 | ||
XM_006719969.1 | XP_006720032.1 | 7 | 1207 | |||
XM_011537498.2 | XP_011535800.1 | 7 | 1550 | |||
XM_011537501.2 | XP_011535803.1 | 7 | 1426 | |||
XM_011537502.2 | XP_011535804.1 | 7 | 1426 | |||
XM_011537504.2 | XP_011535806.1 | 7 | 1426 | |||
XM_011537507.2 | XP_011535809.1 | 7 | 1426 | |||
XM_017020629.1 | XP_016876118.1 | 7 | 1426 | |||
XM_017020631.1 | XP_016876120.1 | 6 | 1234 | |||
XM_017020632.1 | XP_016876121.1 | 6 | 1234 | |||
XM_017020636.1 | XP_016876125.1 | 7 | 1426 | |||
GenBank mRNA | AF209505.1 | - | 6 | 640 | ||
AK000630.1 | - | 7 | 1323 | |||
AK074415.1 | - | 4 | 452 | |||
AY358768.1 | - | 7 | 1204 | |||
BC012564.1 | - | 7 | 1388 | 4080970 | ||
BC068515.1 | - | 7 | 1424 | 4819377 |