Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM118A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104595.1 | NP_001098065.1 | 8, 9 | 1580 | ||
NM_017911.2 | NP_060381.2 | 7, 8 | 1253 | |||
XM_011530247.2 | XP_011528549.1 | 7, 8 | 1371 | |||
XM_011530254.2 | XP_011528556.1 | 7, 8 | 1161 | |||
XM_017028841.1 | XP_016884330.1 | 7, 8 | 1156 | |||
XM_017028842.1 | XP_016884331.1 | 6, 7 | 923 | |||
XM_017028843.1 | XP_016884332.1 | 6, 7 | 920 | |||
XM_017028845.1 | XP_016884334.1 | 8, 9 | 1356 | |||
XM_017028846.1 | XP_016884335.1 | 7, 8 | 1240 | |||
XM_017028847.1 | XP_016884336.1 | 7, 8 | 1884 | |||
XM_017028848.1 | XP_016884337.1 | 7, 8 | 1282 | |||
XM_017028849.1 | XP_016884338.1 | 7, 8 | 1118 | |||
XM_017028850.1 | XP_016884339.1 | 7, 8 | 1241 | |||
XM_017028851.1 | XP_016884340.1 | 7, 8 | 1872 | |||
XM_017028852.1 | XP_016884341.1 | 7, 8 | 1278 | |||
XM_017028854.1 | XP_016884343.1 | 6, 7 | 1005 | |||
XM_017028861.1 | XP_016884350.1 | 7, 8 | 1555 | |||
GenBank mRNA | AK000642.1 | - | 7, 8 | 1223 | ||
AK125027.1 | - | 8, 9 | 1798 | |||
AK302205.1 | - | 4, 5 | 732 | |||
BC013696.1 | - | 7, 8 | 1010 | 3860694 | ||
CB961784.1 | - | 4, 5 | 543 | |||
CR456403.1 | - | 6, 7 | 979 |