Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017923.3 | NP_060393.1 | 1 | 178 | ||
GenBank mRNA | AK000675.1 | - | 1 | 67 | ||
AK301408.1 | - | 1 | 67 | |||
AW118953.1 | - | 1 | 494 |