Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017931.2 | NP_060401.2 | 8 | 860 | ||
XM_011530259.1 | XP_011528561.1 | 9 | 988 | |||
GenBank mRNA | AK000706.1 | - | 8 | 816 | ||
AK300606.1 | - | 7 | 666 | |||
BC018918.2 | - | 8 | 801 | 4130906 |