Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100818.1 | NP_001094288.1 | 3 | 642 | ||
NM_017933.4 | NP_060403.3 | 4 | 853 | |||
XM_011511384.2 | XP_011509686.1 | 3 | 525 | |||
XM_017004404.1 | XP_016859893.1 | 4 | 5784 | |||
XM_017004405.1 | XP_016859894.1 | 5 | 723 | |||
XM_017004406.1 | XP_016859895.1 | 3 | 381 | |||
XM_017004407.1 | XP_016859896.1 | 2 | 438 | |||
GenBank mRNA | AB075874.1 | - | 4 | 508 | ||
AK000708.1 | - | 3 | 628 | |||
AK096636.1 | - | 3 | 584 | |||
AK125359.1 | - | 4 | 777 | |||
AY317148.1 | - | 3 | 474 | |||
BC040164.1 | - | 2 | 429 | 5589172 |