Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 28 | 4759 | ||
NM_017944.3 | NP_060414.3 | 27 | 4555 | |||
XM_005252997.1 | XP_005253054.1 | 29 | 4819 | |||
XM_011520196.1 | XP_011518498.1 | 21 | 3934 | |||
XM_017017953.1 | XP_016873442.1 | 29 | 4875 | |||
XM_017017954.1 | XP_016873443.1 | 21 | 5082 | |||
GenBank mRNA | AK000734.1 | - | 9 | 1270 | ||
AK027362.1 | - | 27 | 3960 | |||
AK074076.1 | - | 13 | 2247 | |||
AK093714.1 | - | 4 | 2281 | |||
AK127264.1 | - | 28 | 4555 | |||
AK225358.1 | - | 9 | 1270 | |||
AL832991.2 | - | 13 | 2170 | |||
BC000226.1 | - | 6 | 526 | |||
BC030562.1 | - | 9 | 1609 | |||
BC047044.1 | - | 9 | 1240 |