Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256394.1 | NP_001243323.1 | 8 | 1375 | ||
NM_017948.5 | NP_060418.4 | 7 | 1271 | |||
NR_046106.1 | 7 | 1271 | ||||
XM_006717166.3 | XP_006717229.1 | 7 | 1137 | |||
XM_006717167.3 | XP_006717230.1 | 7 | 1259 | |||
XM_006717168.3 | XP_006717231.1 | 7 | 1030 | |||
XM_006717169.3 | XP_006717232.1 | 8 | 1290 | |||
XM_006717170.3 | XP_006717233.1 | 8 | 1158 | |||
XM_006717172.3 | XP_006717235.1 | 8 | 1482 | |||
XM_006717173.3 | XP_006717236.1 | 9 | 1595 | |||
XM_011518824.2 | XP_011517126.1 | 6 | 855 | |||
XM_011518827.2 | XP_011517129.1 | 5 | 899 | |||
XM_017014876.1 | XP_016870365.1 | 7 | 1137 | |||
XM_017014877.1 | XP_016870366.1 | 8 | 1158 | |||
XM_017014878.1 | XP_016870367.1 | 8 | 1608 | |||
XM_017014879.1 | XP_016870368.1 | 8 | 1437 | |||
XM_017014880.1 | XP_016870369.1 | 8 | 1237 | |||
GenBank mRNA | AB105104.1 | - | 7 | 1237 | ||
AB109030.1 | - | 6 | 934 | |||
AK001049.1 | - | 7 | 1153 | |||
AK022755.1 | - | 4 | 684 | |||
AK024245.1 | - | 1 | 29 | |||
AK297355.1 | - | 7 | 1154 | |||
AK307408.1 | - | 7 | 1162 | |||
BC064942.1 | - | 1 | 194 | |||
BC146810.1 | - | 8 | 1131 | 8860369 | ||
CR627363.1 | - | 7 | 1015 |