Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOL8 SIRNA Assays ›
Gene Symbol:
NOL8
Gene Name:
nucleolar protein 8
Gene Aliases:
C9orf34, NOP132, bA62C3.3, bA62C3.4
Chromosome Location:
Chr.9: 92297358 - 92325594 on Build GRCh38
UniGene ID:
Hs.442199
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30039
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOL8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256394.1
NP_001243323.1
17, 18
3886
NM_017948.5
NP_060418.4
16, 17
3782
NR_046106.1
16, 17
4576
XM_006717166.3
XP_006717229.1
16, 17
3648
XM_006717167.3
XP_006717230.1
16, 17
3770
XM_006717168.3
XP_006717231.1
15, 16
3427
XM_006717169.3
XP_006717232.1
17, 18
3801
XM_006717170.3
XP_006717233.1
17, 18
3669
XM_006717172.3
XP_006717235.1
17, 18
3993
XM_006717173.3
XP_006717236.1
18, 19
4106
XM_011518824.2
XP_011517126.1
15, 16
3366
XM_011518827.2
XP_011517129.1
14, 15
3410
XM_017014876.1
XP_016870365.1
15, 16
3534
XM_017014877.1
XP_016870366.1
16, 17
3555
XM_017014878.1
XP_016870367.1
17, 18
4119
XM_017014879.1
XP_016870368.1
17, 18
3948
XM_017014880.1
XP_016870369.1
16, 17
3634
GenBank mRNA
AB105104.1
-
16, 17
3748
AB109030.1
-
15, 16
3445
AK000743.1
-
10, 11
1824
AK024245.1
-
10, 11
2540
AK024786.1
-
10, 11
1211
AK225322.1
-
10, 11
1823
BC013788.1
-
10, 11
2030
BC146810.1
-
17, 18
3642
8860369
CR627363.1
-
16, 17
4320
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
rRNA processing
positive regulation of cell growth
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline