Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017952.5 | NP_060422.4 | 12 | 972 | ||
GenBank mRNA | AK000765.1 | - | 2 | 139 | ||
AK293962.1 | - | 11 | 791 | |||
AK315676.1 | - | 6 | 426 | |||
BC001758.1 | - | 1 | 46 | |||
BC011832.2 | - | 12 | 907 | 3939273 |