Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277.2 | NP_001268.2 | 5, 6 | 981 | ||
NM_212469.1 | NP_997634.1 | 4, 5 | 927 | |||
XM_017017147.1 | XP_016872636.1 | 4, 5 | 865 | |||
XM_017017148.1 | XP_016872637.1 | 4, 5 | 2337 | |||
GenBank mRNA | BC036471.2 | - | 4, 5 | 805 | 5265994 | |
D10704.1 | - | 5, 6 | 791 |