Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 14 | 1608 | ||
NM_001190267.1 | NP_001177196.1 | 14 | 1606 | |||
NM_017974.3 | NP_060444.3 | 13 | 1553 | |||
NM_030803.6 | NP_110430.5 | 14 | 1610 | |||
NM_198890.2 | NP_942593.2 | 10 | 1121 | |||
XM_005246082.2 | XP_005246139.1 | 15 | 1598 | |||
XM_005246084.2 | XP_005246141.1 | 12 | 1137 | |||
XM_005246086.2 | XP_005246143.1 | 11 | 1086 | |||
XM_006712608.2 | XP_006712671.1 | 14 | 1409 | |||
GenBank mRNA | AK001690.1 | - | 13 | 1398 | ||
AK024453.1 | - | 12 | 2421 | |||
AK026330.1 | - | 1 | 88 | |||
AK027854.1 | - | 12 | 1135 | |||
AK074434.1 | - | 8 | 580 | |||
AK123876.1 | - | 11 | 972 | |||
AK308400.1 | - | 13 | 1449 | |||
AY358182.1 | - | 13 | 1772 | |||
AY398617.1 | - | 14 | 1499 | |||
BC071846.1 | - | 10 | 959 | |||
BC117337.1 | - | 14 | 1272 | |||
BC143900.1 | - | 13 | 1215 | |||
EF079889.1 | - | 13 | 1296 | |||
EF079890.1 | - | 10 | 864 | |||
EF560734.1 | - | 13 | 1423 | |||
JQ924062.1 | - | 13 | 1296 |