Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017984.4 | NP_060454.3 | 16 | 1800 | ||
XM_006716035.3 | XP_006716098.1 | 16 | 2237 | |||
XM_006716036.2 | XP_006716099.1 | 16 | 1741 | |||
XM_006716037.3 | XP_006716100.1 | 16 | 2234 | |||
XM_006716038.3 | XP_006716101.1 | 16 | 2234 | |||
XM_006716040.3 | XP_006716103.1 | 15 | 2156 | |||
GenBank mRNA | AK302108.1 | - | 15 | 1537 | ||
AL136735.1 | - | 16 | 1671 | |||
BC002725.1 | - | 13 | 1501 | 3631266 | ||
KU178677.1 | - | 11 | 1063 | |||
KU178678.1 | - | 11 | 1066 | |||
KU178679.1 | - | 11 | 1063 |