Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310332.1 | NP_001297261.1 | 15 | 2459 | ||
NM_017999.4 | NP_060469.4 | 15 | 2803 | |||
GenBank mRNA | AB265810.1 | - | 15 | 2554 | ||
AK000973.1 | - | 8 | 1052 | |||
AK027154.1 | - | 7 | 1196 | |||
AK055542.1 | - | 9 | 1600 | |||
AK122711.1 | - | 14 | 2088 | |||
AK225832.1 | - | 15 | 2753 | |||
AK291247.1 | - | 15 | 2755 | |||
AY256461.1 | - | 15 | 2447 | |||
BC009821.2 | - | 8 | 1157 | |||
BC012077.1 | - | 15 | 2723 | 4653017 | ||
BC017376.2 | - | 8 | 1147 |