Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008224.1 | NP_001008225.1 | 16 | 3749 | ||
NM_018003.2 | NP_060473.2 | 16 | 3526 | |||
XM_005254529.4 | XP_005254586.1 | 15 | 3743 | |||
XM_011521752.2 | XP_011520054.2 | 16 | 3855 | |||
XM_011521753.2 | XP_011520055.2 | 15 | 3823 | |||
XM_011521754.2 | XP_011520056.1 | 15 | 3304 | |||
XM_017022394.1 | XP_016877883.1 | 15 | 3575 | |||
XM_017022395.1 | XP_016877884.1 | 16 | 3386 | |||
GenBank mRNA | AB046781.2 | - | 16 | 3422 | ||
AF322916.1 | - | 16 | 3471 | |||
AK000990.1 | - | 1 | 1487 | |||
BC113407.1 | - | 16 | 3445 | 8322459 | ||
BC113409.1 | - | 16 | 3445 | 8322461 | ||
BC143266.1 | - | 16 | 3445 | 9051768 | ||
BC143267.1 | - | 13 | 3118 | 9051769 |