Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 12 | 3891 | ||
GenBank mRNA | AK128806.1 | - | 3 | 1384 | ||
AY923834.1 | - | 12 | 3892 | |||
BC035896.1 | - | 9 | 2412 |