Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143824.1 | NP_001137296.1 | 2, 3 | 332 | ||
NM_018018.4 | NP_060488.2 | 3, 4 | 524 | |||
XM_005268997.2 | XP_005269054.1 | 2, 3 | 333 | |||
GenBank mRNA | AB055003.1 | - | 1, 2 | 156 | ||
AF193836.1 | - | 2, 3 | 312 | |||
AF305814.1 | - | 3, 4 | 489 | |||
AK001053.1 | - | 3, 4 | 400 | |||
AK291168.1 | - | 2, 3 | 333 | |||
BC069819.1 | - | 2, 3 | 385 | 7262399 | ||
BC101827.1 | - | 1, 2 | 169 | 8069333 | ||
BC104913.1 | - | 1, 2 | 168 | 8143916 | ||
DB139964.1 | - | 2, 3 | 332 |