Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018025.2 | NP_060495.2 | 5 | 788 | ||
NR_135270.1 | 5 | 788 | ||||
XM_006723255.3 | XP_006723318.1 | 5 | 503 | |||
GenBank mRNA | AF434677.1 | - | 5 | 474 | ||
AK096005.1 | - | 5 | 482 | |||
AK292503.1 | - | 5 | 482 | |||
AK315712.1 | - | 5 | 500 | |||
AL834465.1 | - | 5 | 479 | |||
BC006108.1 | - | 5 | 788 | 3029084 |