Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143888.2 | NP_001137360.1 | 5 | 499 | ||
NM_001143889.2 | NP_001137361.1 | 4 | 280 | |||
NM_001143890.2 | NP_001137362.1 | 3 | 163 | |||
NM_001300958.1 | NP_001287887.1 | 5 | 499 | |||
NM_018045.7 | NP_060515.3 | 5 | 448 | |||
NR_125352.1 | 5 | 448 | ||||
XM_005270986.4 | XP_005271043.1 | 5 | 428 | |||
XM_011541677.2 | XP_011539979.1 | 5 | 746 | |||
GenBank mRNA | AK001138.1 | - | 5 | 418 | ||
AK021440.1 | - | 5 | 427 | |||
AK225026.1 | - | 5 | 429 | |||
AK225717.1 | - | 5 | 418 | |||
AK297352.1 | - | 5 | 427 | |||
AK297604.1 | - | 4 | 249 | |||
AK300231.1 | - | 5 | 357 | |||
AK300306.1 | - | 5 | 468 | |||
AK304477.1 | - | 3 | 143 | |||
AK316172.1 | - | 4 | 261 | |||
BC028381.2 | - | 5 | 420 | 4812029 | ||
BP363955.1 | - | 5 | 448 | |||
BX641056.1 | - | 4 | 465 | |||
CR457252.1 | - | 4 | 401 | |||
CR749228.1 | - | 4 | 351 |