Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018051.4 | NP_060521.4 | 14 | 1887 | ||
XM_005249549.1 | XP_005249606.1 | 11 | 1479 | |||
XM_006716041.2 | XP_006716104.1 | 14 | 1911 | |||
XM_011516367.1 | XP_011514669.1 | 14 | 1753 | |||
XM_011516368.2 | XP_011514670.1 | 14 | 1911 | |||
XM_011516369.2 | XP_011514671.1 | 14 | 1742 | |||
XM_017012381.1 | XP_016867870.1 | 14 | 1933 | |||
XM_017012382.1 | XP_016867871.1 | 14 | 1902 | |||
XM_017012383.1 | XP_016867872.1 | 14 | 1963 | |||
XM_017012384.1 | XP_016867873.1 | 11 | 1470 | |||
XM_017012385.1 | XP_016867874.1 | 13 | 1644 | |||
XM_017012386.1 | XP_016867875.1 | 11 | 1423 | |||
XM_017012387.1 | XP_016867876.1 | 14 | 1763 | |||
XM_017012388.1 | XP_016867877.1 | 4 | 647 | |||
GenBank mRNA | AK027228.1 | - | 3 | 244 | ||
AL137660.1 | - | 10 | 1060 | |||
BC014491.1 | - | 14 | 1887 | 4906683 | ||
CR627036.1 | - | 9 | 3529 |