Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114636.1 | NP_001108108.1 | 4 | 320 | ||
NM_018062.3 | NP_060532.2 | 4 | 320 | |||
XM_005264395.3 | XP_005264452.1 | 4 | 320 | |||
XM_011532939.2 | XP_011531241.1 | 4 | 320 | |||
XM_011532940.2 | XP_011531242.1 | 4 | 320 | |||
XM_011532941.1 | XP_011531243.1 | 4 | 320 | |||
XM_011532942.2 | XP_011531244.1 | 4 | 320 | |||
XM_017004414.1 | XP_016859903.1 | 4 | 320 | |||
XM_017004415.1 | XP_016859904.1 | 4 | 320 | |||
GenBank mRNA | AK001197.1 | - | 4 | 286 | ||
AK225147.1 | - | 4 | 263 | |||
AK297736.1 | - | 4 | 295 | |||
AL036481.2 | - | 4 | 334 | |||
BC009042.1 | - | 4 | 290 | 3870828 | ||
BC054517.1 | - | 4 | 295 | 5180184 | ||
DB032206.1 | - | 4 | 320 |