Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114636.1 | NP_001108108.1 | 13 | 1162 | ||
NM_018062.3 | NP_060532.2 | 13 | 1147 | |||
XM_005264395.3 | XP_005264452.1 | 13 | 1147 | |||
XM_005264397.4 | XP_005264454.1 | 9 | 866 | |||
XM_011532939.2 | XP_011531241.1 | 14 | 1207 | |||
XM_011532940.2 | XP_011531242.1 | 14 | 1192 | |||
XM_011532941.1 | XP_011531243.1 | 14 | 1207 | |||
XM_011532942.2 | XP_011531244.1 | 13 | 1162 | |||
XM_011532944.2 | XP_011531246.1 | 10 | 884 | |||
XM_011532945.2 | XP_011531247.1 | 9 | 852 | |||
XM_017004414.1 | XP_016859903.1 | 14 | 1192 | |||
XM_017004416.1 | XP_016859905.1 | 9 | 866 | |||
GenBank mRNA | AK001197.1 | - | 13 | 1113 | ||
AK225147.1 | - | 12 | 1003 | |||
BC009042.1 | - | 13 | 1117 | 3870828 | ||
BC054517.1 | - | 13 | 1122 | 5180184 |