Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018064.3 | NP_060534.1 | 2 | 886 | ||
GenBank mRNA | AK001204.1 | - | 2 | 886 | ||
AK222701.1 | - | 2 | 851 | |||
BC000764.2 | - | 2 | 827 | 2822886 | ||
BC003042.1 | - | 2 | 827 | 2822886 | ||
BC005051.1 | - | 2 | 564 | 2820292 |