Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304496.1 | NP_001291425.1 | 6 | 1012 | ||
NM_018073.7 | NP_060543.5 | 7 | 1495 | |||
GenBank mRNA | AF360739.1 | - | 7 | 1448 | ||
AF439153.1 | - | 6 | 1205 | |||
AK001231.1 | - | 4 | 700 | |||
AK022923.1 | - | 4 | 665 | |||
AK056523.1 | - | 7 | 1457 | |||
AK291166.1 | - | 7 | 1407 | |||
AK301120.1 | - | 5 | 727 | |||
AL832730.1 | - | 7 | 1445 | |||
BC075058.2 | - | 6 | 1229 | 30915367 | ||
BC109063.2 | - | 7 | 1414 | 40033795 |