Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166135.1 | NP_001159607.1 | 10 | 1131 | ||
NM_018077.2 | NP_060547.2 | 14 | 1554 | |||
XM_011516370.2 | XP_011514672.1 | 15 | 1661 | |||
XM_017012389.1 | XP_016867878.1 | 15 | 1662 | |||
XM_017012390.1 | XP_016867879.1 | 14 | 1554 | |||
GenBank mRNA | AK001239.1 | - | 14 | 1554 | ||
AK222716.1 | - | 14 | 1516 | |||
AK300500.1 | - | 10 | 1093 | |||
BC013889.2 | - | 14 | 1508 | 3926178 | ||
KU178685.1 | - | 14 | 1439 |