Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018079.4 | NP_060549.4 | 4 | 591 | ||
XM_011532946.1 | XP_011531248.1 | 4 | 592 | |||
XM_017004417.1 | XP_016859906.1 | 4 | 592 | |||
GenBank mRNA | AK056536.1 | - | 4 | 561 |