Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284220.1 | NP_001271149.1 | 12 | 1459 | ||
NM_001284222.1 | NP_001271151.1 | 13 | 1494 | |||
NM_001284224.1 | NP_001271153.1 | 11 | 1356 | |||
NM_001284225.1 | NP_001271154.1 | 11 | 1395 | |||
NM_001284226.1 | NP_001271155.1 | 12 | 1420 | |||
NM_018091.5 | NP_060561.3 | 13 | 1559 | |||
XM_006716354.2 | XP_006716417.1 | 12 | 1449 | |||
XM_017013604.1 | XP_016869093.1 | 13 | 1559 | |||
GenBank mRNA | AK022626.1 | - | 13 | 1515 | ||
AK223047.1 | - | 12 | 1345 | |||
AK293424.1 | - | 11 | 1322 | |||
AK295574.1 | - | 12 | 1348 | |||
AK296475.1 | - | 12 | 1414 | |||
AK298265.1 | - | 12 | 1310 | |||
AK302140.1 | - | 11 | 1321 | |||
AK315985.1 | - | 11 | 1299 | |||
AK316053.1 | - | 12 | 1363 | |||
AL834273.1 | - | 13 | 1417 | |||
BC001240.1 | - | 13 | 1449 | 3457039 | ||
BX648011.1 | - | 13 | 1493 |