Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134387.1 | NP_001127859.1 | 3 | 523 | ||
NM_001134388.1 | NP_001127860.1 | 4 | 432 | |||
NM_001134389.1 | NP_001127861.1 | 3 | 301 | |||
NM_018106.3 | NP_060576.1 | 3 | 366 | |||
XM_005249793.2 | XP_005249850.1 | 4 | 476 | |||
XM_005249794.3 | XP_005249851.1 | 3 | 466 | |||
XM_005249795.2 | XP_005249852.1 | 3 | 299 | |||
XM_005249796.3 | XP_005249853.1 | 3 | 413 | |||
GenBank mRNA | AF201931.1 | - | 2 | 295 | ||
AK001341.1 | - | 3 | 282 | |||
AK223532.1 | - | 3 | 366 | |||
AK308971.1 | - | 3 | 281 | |||
AL136674.1 | - | 3 | 274 | |||
AY359090.1 | - | 3 | 454 | |||
BC001239.1 | - | 4 | 385 | 3459274 | ||
BQ925460.1 | - | 4 | 392 | |||
CN365375.1 | - | 3 | 482 | |||
CR533511.1 | - | 1 | 74 |