Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288983.1 | NP_001275912.1 | 3 | 384 | ||
NM_001288984.1 | NP_001275913.1 | 3 | 459 | |||
NM_018115.3 | NP_060585.2 | 3 | 384 | |||
XM_005263102.4 | XP_005263159.1 | 3 | 440 | |||
XM_005263105.4 | XP_005263162.1 | 3 | 498 | |||
XM_017008354.1 | XP_016863843.1 | 3 | 360 | |||
XM_017008355.1 | XP_016863844.1 | 2 | 248 | |||
GenBank mRNA | AK001360.1 | - | 2 | 119 | ||
AK022683.1 | - | 1 | 38 | |||
AK300074.1 | - | 3 | 360 | |||
AK307540.1 | - | 3 | 367 | |||
BC048351.1 | - | 2 | 116 | |||
BC054040.1 | - | 3 | 325 | |||
BC063797.1 | - | 3 | 355 | |||
BC107895.1 | - | 3 | 364 | |||
BJ989617.1 | - | 3 | 391 | |||
CR749574.1 | - | 3 | 435 | |||
DB047277.1 | - | 3 | 440 |