Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSTO2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024117.2 | 3 | 320 | |||
GenBank mRNA | AK293928.1 | - | 3 | 320 | ||
AK295146.1 | - | 3 | 320 | |||
BC099909.1 | - | 3 | 270 | |||
DC387604.1 | - | 3 | 320 |