Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018129.3 | NP_060599.1 | 5 | 680 | ||
XM_005257500.3 | XP_005257557.1 | 5 | 870 | |||
XM_011524968.2 | XP_011523270.1 | 4 | 453 | |||
XM_017024813.1 | XP_016880302.1 | 5 | 468 | |||
GenBank mRNA | AF468030.1 | - | 5 | 527 | ||
AK001397.1 | - | 5 | 632 | |||
AK223242.1 | - | 5 | 611 | |||
AK303536.1 | - | 4 | 455 | |||
AK303792.1 | - | 4 | 579 | |||
BC006525.2 | - | 5 | 567 | 2966366 | ||
FJ224333.1 | - | 5 | 527 |