Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127182.1 | NP_001120654.1 | 6 | 1164 | ||
NM_018131.4 | NP_060601.3 | 6 | 1286 | |||
XM_011539918.1 | XP_011538220.1 | 6 | 1300 | |||
XM_011539919.1 | XP_011538221.1 | 6 | 1168 | |||
XM_011539920.2 | XP_011538222.1 | 6 | 994 | |||
XM_017016372.1 | XP_016871861.1 | 6 | 1140 | |||
XM_017016373.1 | XP_016871862.1 | 6 | 1162 | |||
GenBank mRNA | AB091343.1 | - | 6 | 1275 | ||
AK001402.1 | - | 5 | 886 | |||
AK223495.1 | - | 6 | 1267 | |||
AK315536.1 | - | 6 | 1283 | |||
AY788918.1 | - | 6 | 1286 | |||
BC008947.2 | - | 6 | 1266 | 3028566 | ||
BX640718.1 | - | 7 | 1206 |