Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193343.1 | NP_001180272.1 | 3, 4 | 281 | ||
NM_018135.3 | NP_060605.1 | 3, 4 | 281 | |||
XM_006715134.2 | XP_006715197.1 | 3, 4 | 257 | |||
GenBank mRNA | AB049952.1 | - | 3, 4 | 258 | ||
AK001410.1 | - | 3, 4 | 287 | |||
AK300757.1 | - | 3, 4 | 252 | |||
BC010364.1 | - | 3, 4 | 256 | 4248325 | ||
DQ884400.1 | - | 3, 4 | 248 |