Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134415.1 | NP_001127887.1 | 6 | 1875 | ||
NM_001286749.1 | NP_001273678.1 | 5 | 1496 | |||
NM_018138.4 | NP_060608.1 | 6 | 1526 | |||
GenBank mRNA | AK001422.1 | - | 6 | 1499 | ||
AK124362.1 | - | 5 | 1271 | |||
BC025748.1 | - | 6 | 1512 | 5200082 |