Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018141.3 | NP_060611.2 | 4, 5 | 362 | ||
XM_005249206.2 | XP_005249263.1 | 4, 5 | 371 | |||
XM_011514724.2 | XP_011513026.1 | 5, 6 | 366 | |||
XM_011514725.2 | XP_011513027.1 | 5, 6 | 358 | |||
XM_017010996.1 | XP_016866485.1 | 4, 5 | 363 | |||
GenBank mRNA | AF113220.1 | - | 4, 5 | 322 | ||
AK001429.1 | - | 4, 5 | 331 | |||
AK098446.1 | - | 4, 5 | 325 | |||
AK298219.1 | - | 4, 5 | 345 | |||
AK316599.1 | - | 4, 5 | 328 | |||
BC012560.1 | - | 4, 5 | 332 | 4277803 | ||
BG776014.1 | - | 4, 5 | 364 |