Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018141.3 | NP_060611.2 | 3 | 186 | ||
XM_005249206.2 | XP_005249263.1 | 3 | 195 | |||
XM_011514724.2 | XP_011513026.1 | 4 | 190 | |||
XM_011514725.2 | XP_011513027.1 | 4 | 182 | |||
XM_017010996.1 | XP_016866485.1 | 3 | 187 | |||
GenBank mRNA | AF113220.1 | - | 3 | 146 | ||
AF277180.1 | - | 3 | 148 | |||
AK001429.1 | - | 3 | 155 | |||
AK098446.1 | - | 3 | 149 | |||
AK316599.1 | - | 3 | 152 | |||
BC012560.1 | - | 3 | 156 | 4277803 | ||
BG776014.1 | - | 3 | 187 |