Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018142.2 | NP_060612.2 | 13 | 1741 | ||
XM_005273555.3 | XP_005273612.1 | 13 | 2002 | |||
XM_005273556.3 | XP_005273613.1 | 13 | 1783 | |||
XM_005273557.3 | XP_005273614.1 | 13 | 1786 | |||
XM_006716355.2 | XP_006716418.1 | 13 | 1891 | |||
XM_017013606.1 | XP_016869095.1 | 13 | 1786 | |||
XM_017013607.1 | XP_016869096.1 | 13 | 2002 | |||
XM_017013608.1 | XP_016869097.1 | 12 | 1394 | |||
XM_017013609.1 | XP_016869098.1 | 12 | 1718 | |||
GenBank mRNA | AK001431.1 | - | 13 | 1661 | ||
AL442078.1 | - | 4 | 482 | |||
BC001915.1 | - | 11 | 1378 | |||
BC006209.2 | - | 11 | 1378 | |||
BC051282.1 | - | 11 | 1392 | |||
CR457262.1 | - | 10 | 1271 |