Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142627.2 | NP_001136099.1 | 10 | 1151 | ||
NM_001142628.1 | NP_001136100.1 | 9 | 1085 | |||
NM_018144.3 | NP_060614.2 | 10 | 1151 | |||
NR_024576.2 | 10 | 1151 | ||||
NR_024577.2 | 11 | 1284 | ||||
GenBank mRNA | AF346603.1 | - | 10 | 1092 | ||
AK001440.1 | - | 3 | 386 | |||
AK022027.1 | - | 4 | 534 | |||
AK022640.1 | - | 10 | 1086 | |||
AK057532.1 | - | 11 | 1225 | |||
AK292295.1 | - | 10 | 1101 | |||
AK301841.1 | - | 9 | 1038 | |||
BC026179.1 | - | 10 | 1101 | 4823130 |
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077032.1 | - | 10 | 1094 |