Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040616.2 | NP_001035706.1 | 7 | 2152 | ||
XM_005254941.1 | XP_005254998.1 | 7 | 2100 | |||
XM_005254943.1 | XP_005255000.1 | 6 | 1924 | |||
XM_017022398.1 | XP_016877887.1 | 7 | 2194 | |||
XM_017022399.1 | XP_016877888.1 | 5 | 1569 | |||
XM_017022400.1 | XP_016877889.1 | 8 | 2410 | |||
GenBank mRNA | AB083157.1 | - | 6 | 1982 | ||
AK095448.1 | - | 2 | 2202 |