Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104647.1 | NP_001098117.1 | 6 | 910 | ||
NM_018155.2 | NP_060625.2 | 6 | 910 | |||
XM_005247575.4 | XP_005247632.1 | 3 | 2021 | |||
GenBank mRNA | AK001480.1 | - | 6 | 884 | ||
AK296785.1 | - | 5 | 751 | |||
BC014064.1 | - | 6 | 843 | 3842022 | ||
BC019859.1 | - | 7 | 872 | |||
BC036272.1 | - | 7 | 872 |