Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018157.2 | NP_060627.2 | 3 | 589 | ||
XM_005268998.3 | XP_005269055.1 | 3 | 615 | |||
XM_011538526.2 | XP_011536828.1 | 2 | 971 | |||
XM_011538527.2 | XP_011536829.1 | 2 | 877 | |||
XM_011538528.2 | XP_011536830.1 | 2 | 973 | |||
XM_011538529.2 | XP_011536831.1 | 2 | 968 | |||
XM_017019548.1 | XP_016875037.1 | 3 | 615 | |||
XM_017019549.1 | XP_016875038.1 | 2 | 480 | |||
GenBank mRNA | AK001482.1 | - | 3 | 496 | ||
AY221116.1 | - | 3 | 398 | |||
BC034220.1 | - | 4 | 632 | 5264088 | ||
BC132689.1 | - | 3 | 570 | 40146711 | ||
BC144056.1 | - | 3 | 570 | 9052576 | ||
DA534432.1 | - | 1 | 114 |