Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283056.1 | NP_001269985.1 | 1, 2 | 377 | ||
NM_001283057.1 | NP_001269986.1 | 1, 2 | 377 | |||
NM_001283058.1 | NP_001269987.1 | 1, 2 | 377 | |||
NM_018168.3 | NP_060638.2 | 1, 2 | 377 | |||
XM_005267806.1 | XP_005267863.1 | 1, 2 | 370 | |||
XM_005267813.1 | XP_005267870.1 | 1, 2 | 366 | |||
XM_005267814.1 | XP_005267871.1 | 1, 2 | 366 | |||
XM_006720188.2 | XP_006720251.1 | 1, 2 | 370 | |||
GenBank mRNA | AK001512.1 | - | 1, 2 | 366 | ||
AK223127.1 | - | 1, 2 | 365 | |||
AK307238.1 | - | 1, 2 | 360 | |||
BC117405.1 | - | 1, 2 | 271 | 40125956 | ||
BC143565.1 | - | 1, 2 | 271 | |||
BC143566.1 | - | 1, 2 | 271 | 9052077 | ||
BC143567.1 | - | 1, 2 | 271 | 9052078 | ||
BX470593.1 | - | 1, 2 | 389 |