Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 4 | 352 | ||
NM_018172.2 | NP_060642.2 | 4 | 373 | |||
NM_152563.2 | NP_689776.1 | 3 | 271 | |||
GenBank mRNA | AK001523.1 | - | 4 | 347 | ||
AK130709.1 | - | 3 | 268 | |||
AK293221.1 | - | 3 | 245 | |||
AK311428.1 | - | 4 | 329 | |||
AK311518.1 | - | 4 | 352 | |||
AK311675.1 | - | 5 | 458 | |||
AK315454.1 | - | 3 | 245 | |||
BC032519.1 | - | 3 | 271 | 5494088 | ||
BQ921333.1 | - | 4 | 322 | |||
DC312827.1 | - | 5 | 457 |