Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF532 SIRNA Assays ›
Gene Symbol:
ZNF532
Gene Name:
zinc finger protein 532
Gene Aliases:
-
Chromosome Location:
Chr.18: 58862600 - 58986480 on Build GRCh38
UniGene ID:
Hs.529023
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30433
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF532
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318726.1
NP_001305655.1
3
2949
NM_001318727.1
NP_001305656.1
3
2507
NM_001318728.1
NP_001305657.1
4
2609
NM_018181.5
NP_060651.2
4
3031
XM_011526054.2
XP_011524356.1
3
2756
XM_011526067.2
XP_011524369.1
1
2272
XM_017025809.1
XP_016881298.1
1
2272
XM_017025810.1
XP_016881299.1
6
3289
XM_017025811.1
XP_016881300.1
4
3352
XM_017025812.1
XP_016881301.1
4
2589
XM_017025813.1
XP_016881302.1
3
2548
XM_017025814.1
XP_016881303.1
3
2503
XM_017025815.1
XP_016881304.1
2
2839
XM_017025816.1
XP_016881305.1
4
2636
XM_017025817.1
XP_016881306.1
4
2837
XM_017025818.1
XP_016881307.1
3
2607
XM_017025819.1
XP_016881308.1
4
2660
XM_017025820.1
XP_016881309.1
3
2756
XM_017025821.1
XP_016881310.1
3
2949
XM_017025822.1
XP_016881311.1
3
2757
XM_017025823.1
XP_016881312.1
2
2514
XM_017025824.1
XP_016881313.1
2
2516
XM_017025825.1
XP_016881314.1
3
3156
XM_017025826.1
XP_016881315.1
3
2757
XM_017025827.1
XP_016881316.1
3
2578
XM_017025828.1
XP_016881317.1
3
2757
XM_017025829.1
XP_016881318.1
2
2515
XM_017025830.1
XP_016881319.1
2
2517
XM_017025831.1
XP_016881320.1
3
2757
GenBank mRNA
AB046849.2
-
3
2467
AF295077.1
-
4
2826
AK001559.1
-
1
217
AK128058.1
-
4
3033
AY039256.1
-
3
3144
BC036366.1
-
4
2519
4828639
BC130618.1
-
3
2601
40146649
BC130620.1
-
4
2683
40146651
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0