Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018190.3 | NP_060660.2 | 18 | 2108 | ||
NM_176824.2 | NP_789794.1 | 18 | 2108 | |||
XM_005263106.3 | XP_005263163.1 | 18 | 2134 | |||
XM_011532079.2 | XP_011530381.1 | 18 | 2179 | |||
XM_011532080.2 | XP_011530382.1 | 18 | 2176 | |||
XM_011532081.2 | XP_011530383.1 | 17 | 2014 | |||
XM_017008357.1 | XP_016863846.1 | 17 | 1966 | |||
XM_017008358.1 | XP_016863847.1 | 17 | 1969 | |||
GenBank mRNA | AF521643.1 | - | 18 | 2072 | ||
AF521644.1 | - | 18 | 2072 | |||
AK001577.1 | - | 18 | 2072 | |||
AK308076.1 | - | 18 | 2097 | |||
BC032691.1 | - | 18 | 2025 | 5240398 |