Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 13 | 1346 | ||
NM_018193.2 | NP_060663.2 | 13 | 1346 | |||
XM_011521756.2 | XP_011520058.1 | 13 | 1370 | |||
XM_011521757.2 | XP_011520059.1 | 13 | 1366 | |||
XM_011521764.2 | XP_011520066.1 | 13 | 1370 | |||
XM_011521767.2 | XP_011520069.1 | 13 | 1340 | |||
GenBank mRNA | AK001581.1 | - | 7 | 735 | ||
AK027564.1 | - | 2 | 233 | |||
AK055176.1 | - | 13 | 1305 | |||
BC140769.1 | - | 13 | 1303 | 9021637 | ||
BC144483.1 | - | 13 | 1303 | 9053010 | ||
EF469766.1 | - | 12 | 1256 | |||
EF567077.1 | - | 12 | 1256 |