Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297760.1 | NP_001284689.1 | 6 | 1321 | ||
NM_001297761.1 | NP_001284690.1 | 5 | 1061 | |||
NM_001297762.1 | NP_001284691.1 | 5 | 1198 | |||
NM_018208.3 | NP_060678.2 | 6 | 1321 | |||
XM_011509714.2 | XP_011508016.1 | 5 | 561 | |||
XM_011509715.1 | XP_011508017.1 | 5 | 866 | |||
XM_017001632.1 | XP_016857121.1 | 5 | 649 | |||
GenBank mRNA | AB073608.1 | - | 6 | 921 | ||
AK001623.1 | - | 6 | 1033 | |||
AK293426.1 | - | 5 | 769 | |||
AK302145.1 | - | 5 | 798 | |||
AK316493.1 | - | 5 | 557 | |||
BC010082.2 | - | 6 | 1142 | 4329734 |