Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 3, 4 | 522 | ||
NM_024662.2 | NP_078938.2 | 5, 6 | 737 | |||
GenBank mRNA | AB051496.1 | - | 5, 6 | 658 | ||
AK001636.1 | - | 5, 6 | 592 | |||
AK022241.1 | - | 5, 6 | 699 | |||
AK074430.1 | - | 1315 | ||||
AK225341.1 | - | 5, 6 | 699 | |||
AK294044.1 | - | 3, 4 | 421 | |||
AL136882.1 | - | 5, 6 | 594 | |||
BC020205.1 | - | 5, 6 | 559 | |||
BC035558.1 | - | 5, 6 | 671 | 5166101 |