Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 6 | 793 | ||
NM_024662.2 | NP_078938.2 | 8 | 1008 | |||
GenBank mRNA | AB051496.1 | - | 8 | 929 | ||
AF489535.1 | - | 4 | 305 | |||
AK001636.1 | - | 8 | 863 | |||
AK022241.1 | - | 8 | 970 | |||
AK074430.1 | - | 1586 | ||||
AK225341.1 | - | 8 | 970 | |||
AK294044.1 | - | 6 | 692 | |||
AL136882.1 | - | 8 | 865 | |||
BC020205.1 | - | 8 | 830 | |||
BC035558.1 | - | 8 | 942 | 5166101 |