Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018214.4 | NP_060684.4 | 7 | 865 | ||
XM_011514726.2 | XP_011513028.1 | 8 | 1229 | |||
XM_011514727.2 | XP_011513029.1 | 6 | 655 | |||
XM_017010997.1 | XP_016866486.1 | 7 | 1157 | |||
GenBank mRNA | AF332199.1 | - | 7 | 812 | ||
AF359380.1 | - | 7 | 588 | |||
AK001637.1 | - | 7 | 569 | |||
AK021896.1 | - | 8 | 1225 | |||
AK292248.1 | - | 7 | 710 | |||
BC003193.1 | - | 7 | 865 | 3504090 |